Aluminiumammoniumsulfat Dodecahydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Aluminium ammonium bis(sulphate) dodecahydrate, Ammonium alum, Ammonium aluminum sulphate dodecahydrate, Aluminum ammonium sulphatedodecahydrate
CAS No.: 7784-26-1
Molecular Formula: H4AlNO8S2.12H2O
Formula Weight: 453.33 (237.14anhy)
Physical Form: Crystalline
Density: 1.65
Appearance: White
Melting Point: 93-94°
Solubility: Soluble in water, glycerol. Insoluble in alcohol
Merck Reference: 14,327
MDL No.: MFCD00051042
Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper plating
Formel:
AlNH₄(SO₄)₂·12H₂O Siedepunkt: 200 °C (1013 hPa) Schmelzpunkt: 93…94 °C Dichte: 1.65 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051042 CAS-Nummer: 7784-26-1 EINECS: 232-055-3 Merck Index: 14,327 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...