Indium(III)oxid ≥99,997% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific

Synonyme: Indium trioxide, Diindium trioxide

10709.09 10709.18
10709.09EA 142 EUR
10709.09 10709.18
Indium(III)oxid ≥99,997% (Metall-Basis), Puratronic®
Indium(III)oxid

Purity: 99.997% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: Powder Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Formel: In₂O₃
Molekulargewicht: 277.64 g/mol
Schmelzpunkt: 850 °C
Dichte: 7.18
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011060
CAS-Nummer: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR