Aluminiumoxid, γ-Phase ≥99%, Nano-Pulver

Lieferant: Thermo Fisher Scientific

44757.14 44757.22 44757.36
44757.14EA 66.5 EUR
44757.14 44757.22 44757.36
Aluminiumoxid, γ-Phase ≥99%, Nano-Pulver
Aluminiumoxid, γ-Phase

Purity: 99+% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 20nm APS Powder, S.A. 100-200m2/g Appearance: White Odour: Odourless Merck Reference: 14,356 MDL No.: MFCD00003424

Formel: Al₂O₃
Siedepunkt: 2980 °C (1013 hPa)
Schmelzpunkt: 2045 °C
Dichte: 3.94 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR