Aluminiumoxid, γ-Phase bimodal, große Fläche Katalysatorträger
Lieferant: Thermo Fisher Scientific
S.A. 255 m2/g, median pore 70 micron and 5000A, total pore volume 1.14cc/g, packing density 0.395 kg/L
Physical Form: 1/8" pellets
Odour: Odourless
S.A. 255m2/g, median pore 70 micron and 5000A, total pore volume 1.14cc/g, packing density 0.395kg/L
Formel:
Al₂O₃ Siedepunkt: 2980 °C Schmelzpunkt: 2045 °C Dichte: 3,965 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...