Aluminiumoxid, γ-Phase ≥99,97% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
39812.14
39812.22
39812.36
39812.14EA
50
EUR
39812.14
39812.22
39812.36
Aluminiumoxid, γ-Phase ≥99,97% (Metall-Basis)
Aluminiumoxid, γ-Phase
Purity: 99.97%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 3 Micron APS Powder, S.A. 80-120m2/g
Odour: Odourless
Merck Reference: 14,356
MDL No.: MFCD00003424
Formel:
Al₂O₃ Siedepunkt: 2980 °C (1013 hPa) Schmelzpunkt: 2045 °C Dichte: 3.94 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003424 CAS-Nummer: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...