Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: ɑ-Tonerde, α-Alumina

42572.22 42572.36 42572.A3
42572.22EA 41.6 EUR
42572.22 42572.36 42572.A3
Aluminiumoxid, α-Phase ≥99,9% (Metall-Basis)
Aluminiumoxid, α-Phase

Purity: 99.9% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 1.0 Micron APS Powder, S.A. 2-4m2/g Refractive Index: 1.765 Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424


Suitable for ceramic, electronic ceramic, and ceramic matrix composites

Formel: Al₂O₃
Siedepunkt: 2980 °C
Schmelzpunkt: 2045 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR