Indium(III)fluorid, wasserfrei ≥96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40118.04
40118.09
40118.04EA
65.5
EUR
40118.04
40118.09
Indium(III)fluorid, wasserfrei ≥96%
Indium(III)fluorid
Purity: 96%
CAS No.: 7783-52-0
Molecular Formula: InF3
Formula Weight: 171.82
Physical Form: Powder
Density: 4.39
Appearance: White
Odour: Odourless
Melting Point: ca 1170°
Boiling Point:
Formel:
F₃In Molekulargewicht: 171.82 g/mol Siedepunkt: >1200 °C Schmelzpunkt: ∼ 1170 °C Dichte: 4.39 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00016149 CAS-Nummer: 7783-52-0 EINECS: 232-005-0 UN-Nummer: 2923 ADR: 8 II Merck Index: 14,4959 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...