Aluminiumfluorid, wasserfrei 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Aluminum fluoride

11497.03 11497.09 11497.18
11497.03EA 25.6 EUR
11497.03 11497.09 11497.18
Aluminiumfluorid, wasserfrei 99+%
Aluminium fluoride

Purity: 99+% CAS No.: 7784-18-1 Molecular Formula: AlF3 Formula Weight: 83.98 Physical Form: Powder Density: 2.882 Melting Point: 1291° subl. Solubility: Practically insoluble in water. Sparingly soluble in acids and alkalies Merck Reference: 14,339 MDL No.: MFCD00003426


In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions

Formel: AlF₃
Molekulargewicht: 83.98 g/mol
Schmelzpunkt: 1291 °C (Subl.)
Dichte: 2.882
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003426
CAS-Nummer: 7784-18-1
EINECS: 232-051-1
UN-Nummer: 3260
ADR: 8 II
Merck Index: 14,339

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR