Aluminiumbromid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Aluminum bromide
Purity: 98%
CAS No.: 7727-15-3
Molecular Formula: AlBr3
Formula Weight: 266.71
Physical Form: Crystalline
Density: 3.21
Appearance: White to orange
Odour: Pungent
Melting Point: 97.5°
Boiling Point: 265°
Solubility: Reacts with water violently, soluble in alcohol, carbon disulfide, ether
Merck Reference: 14,332
UN No.: 1725
MDL No.: MFCD00003421
Alkylation, bromination and isomerization catalyst in organic synthesis
Formel:
AlBr₃ Molekulargewicht: 266.69 g/mol Siedepunkt: 265 °C Schmelzpunkt: 97.5 °C Dichte: 3.21 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003421 CAS-Nummer: 7727-15-3 EINECS: 231-779-7 UN-Nummer: 1725 ADR: 8 II Merck Index: 13,00330 |