Magnesiumsulfat, (Trockenmasse) 98,0-102,0% USP
Lieferant: VWR Chemicals
Synonyme:
MgSO4, Magnesium sulphate anhydrous
7154.1000EA
129
EUR
7154.1000
7154.9025
Magnesiumsulfat, (Trockenmasse) 98,0-102,0% USP
Magnesium sulphate
Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)
Formel:
MgSO₄ Molekulargewicht: 120.37 g/mol Schmelzpunkt: 1127 °C Dichte: 2.65 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011110 CAS-Nummer: 7487-88-9 EINECS: 231-298-2 Merck Index: 12,05731 |
|
Spezifikation Testergebnisse
Assay (by ignition) | 98.0 - 102.0 % |
Identification A | Passes test |
Identification B | Passes test |
Heavy metals (as Pb) | ≤ 10 ppm |
Cl (Chloride) | ≤ 0.014 % |
Fe (Iron) | ≤ 20 μg/g |
Se (Selenium) | ≤ 30 μg/g |
pH (25°C; 50 mg/ml) | 5.0 - 9.2 |
Loss on ignition (450 ± 25°C) | 22.0 - 28.0 % |
Loss on drying | Max. 2 % |
Residues of metal catalysts or reagents | Unlikely by manuf.process |
Residual solvents | Unlikely by manuf.process |
Residual metal catalysts (skip lot): | . |
As (Arsenic) | ≤ 2 ppm |
Cd (Cadmium) | ≤ 1 ppm |
Co (Cobalt) | ≤ 1 ppm |
Hg (Mercury) | ≤ 1 ppm |
Ni (Nickel) | ≤ 1 ppm |
Pb (Lead) | ≤ 2 ppm |
V (Vanadium) | ≤ 1 ppm |
Conforms to USP | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...