Magnesiumoxid ≥95%
Lieferant: Thermo Fisher Scientific
Synonyme:
Magnesia usta
Purity: 95% min
CAS No.: 1309-48-4
Molecular Formula: MgO
Formula Weight: 40.30
Physical Form: -140+325 Mesh Powder
Density: 3.58
Refractive Index: 1.736
Odour: Odourless
Melting Point: 2852°
Boiling Point: 3600°
Solubility: Slightly soluble in water, (solubility increased by CO2). Soluble in acids and ammonium salt solution. Insoluble in alcohol
Merck Reference: 14,5677
MDL No.: MFCD00011109
For plasma spraying, Refractory applications, electrical insulation, pharmaceuticals, cosmetics, paper manufacture, adsorption and catalysis, and in semiconductors, reflector in optical instruments
Formel:
MgO Molekulargewicht: 40.3 g/mol Siedepunkt: 3600 °C Schmelzpunkt: 2852 °C Dichte: 3.58 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011109 CAS-Nummer: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |