Magnesiumfluorid ≥99,99% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

40755.04 40755.09 40755.18
40755.04EA 83 EUR
40755.04 40755.09 40755.18
Magnesiumfluorid ≥99,99% (Metall-Basis), Puratronic®
Magnesium fluoride

Purity: 99.99% CAS No.: 7783-40-6 Molecular Formula: F2Mg Formula Weight: 62.30 Physical Form: Powder Density: 3.148 Refractive Index: 1.35-1.38 Appearance: White Odour: Odourless Melting Point: 1261° Merck Reference: 14,5665 MDL No.: MFCD00011108

Formel: MgF₂
Siedepunkt: 2260 °C (1013 hPa)
Schmelzpunkt: 1261 °C
Dichte: 3.148
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011108
CAS-Nummer: 7783-40-6
EINECS: 231-995-1
Merck Index: 14,5665

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR