Bariumfluorid ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

12338.14 12338.30 12338.A1
12338.14EA 23.9 EUR
12338.14 12338.30 12338.A1
Bariumfluorid ≥99%
Barium fluoride

Purity: 99% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: -40 Mesh Powder Density: 4.83 Refractive Index: 1.4741 Odour: Odourless Melting Point: 1353° Boiling Point: 2260° Solubility: Soluble in HCl, HNO3, HF, and aqueous solutions of ammonium chloride Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450


Ceramic flux, glass manufacture, electronics, and dry-film lubricants

Formel: BaF₂
Siedepunkt: 2260 °C
Schmelzpunkt: 1353 °C
Dichte: 4.83
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003450
CAS-Nummer: 7787-32-8
EINECS: 232-108-0
UN-Nummer: 1564
ADR: 6.1,III
Merck Index: 14,974

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR