Bariumperchlorat, wasserfrei

Lieferant: Thermo Fisher Scientific
Gefahr

11657.22 11657.36
11657.22EA 68.6 EUR
11657.22 11657.36
Bariumperchlorat, wasserfrei
Barium perchlorate

CAS No.: 13465-95-7 Molecular Formula: Ba(ClO4)2 Formula Weight: 336.24 Physical Form: Powder Density: 3.20 Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,987 UN No.: 1447 MDL No.: MFCD00003440


Desiccant, ribonuclease determination

Formel: Ba(ClO₄)₂
Molekulargewicht: 336.23 g/mol
Schmelzpunkt: 505
Dichte: 3.20
Flammpunkt: 21 °C (geschlossener Tiegel)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003440
CAS-Nummer: 13465-95-7
EINECS: 236-710-4
UN-Nummer: 1447
ADR: 5.1 II
Merck Index: 14,987

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR