Kaliumsulfat ≥99,997% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific

10845.06 10845.14
10845.06EA 72.3 EUR
10845.06 10845.14
Kaliumsulfat ≥99,997% (Metall-Basis), Puratronic®
Potassium sulphate

Purity: 99.997% CAS No.: 7778-80-5 Molecular Formula: K2O4S Formula Weight: 174.27 Physical Form: Powder Density: 2.662 Appearance: White Odour: Odourless Melting Point: 1069° Boiling Point: 1689° Merck Reference: 14,7674 MDL No.: MFCD00011388

Formel: K₂SO₄
Siedepunkt: 1689 °C
Schmelzpunkt: 1069 °C
Dichte: 2,662
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011388
CAS-Nummer: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR