Kaliumdihydrogenphosphat ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
A12142.0E A12142.36
A12142.0EEA 156 EUR
A12142.0E A12142.36
Kaliumdihydrogenphosphat ≥98%
Kaliumdihydrogenphosphat
Purity: 98+% CAS No.: 7778-77-0 Molecular Formula: H2KO4P Formula Weight: 136.09 Physical Form: Cryst solid Density: 2.338 Appearance: White Melting Point: 253° Merck Reference: 14,7659 MDL No.: MFCD00011401
Formel: KH₂PO₄
Molekulargewicht: 136.09 g/mol
Siedepunkt: ∼100 °C (1013 hPa)
Schmelzpunkt: 253
Dichte: 2.338
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH-Nummer: 01-2119490224-41
Merck Index: 13,07744