, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
M-Clarity™ Qualitätsstufe = MQ200
P8281-100G
P8281-500G
SIALP8281-100GEA
30.2
EUR
SIALP8281-100G
SIALP8281-500G
, Sigma-Aldrich®
di-Kaliumhydrogenphosphat
Formel:
K₂HPO₄ Molekulargewicht: 174.18 g/mol Schmelzpunkt: 340 Dichte: 2.3 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011383 CAS-Nummer: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...