di-Kaliumhydrogenphosphat

Lieferant: Roth Carl

Synonyme: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

6875.1 6875.2
ROTH6875.1EA 0 EUR
ROTH6875.1 ROTH6875.2
di-Kaliumhydrogenphosphat
di-Kaliumhydrogenphosphat
Formel: K₂HPO₄
Molekulargewicht: 174.18 g/mol
Schmelzpunkt: 340
Dichte: 2.3 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011383
CAS-Nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR