Kaliumtetrahydroborat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Potassium tetrahydridoborate, Potassium borohydride
Purity: 98%
CAS No.: 13762-51-1
Molecular Formula: BH4K
Formula Weight: 53.94
Physical Form: Powder
Density: 1.17
Refractive Index: 1.494
Sensitivity: Moisture Sensitive
Appearance: White
Odour: Odourless
Melting Point:
Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
Formel:
H₄BK Schmelzpunkt: >400 °C (dec.) Dichte: 1.17 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011396 CAS-Nummer: 13762-51-1 EINECS: 237-360-5 UN-Nummer: 1870 ADR: 4.3,I Merck Index: 14,7616 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...