Natriumtetraethylborat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ100

481483-1G 481483-5G
SIAL481483-1GEA 79 EUR
SIAL481483-1G SIAL481483-5G
Natriumtetraethylborat, Sigma-Aldrich®
Sodium tetraethylborate
Formel: C₈H₂₀BNa
MDL Number: MFCD00061547
CAS-Nummer: 15523-24-7
UN-Nummer: 3393
ADR: 4.2,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR