1-Amino-9,10-anthrachinon 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Aminoanthraquinone, 1-Aminoanthracene-9,10-dione

B22901.22 B22901.36
B22901.22EA 40.4 EUR
B22901.22 B22901.36
1-Amino-9,10-anthrachinon 97%
1-Amino-9,10-anthrachinon

Purity: 97% CAS No.: 82-45-1 Molecular Formula: C14H9NO2 Formula Weight: 223.23 Melting Point: 252-255° Solubility: Soluble in alcohol, benzene, chlroform, ether, glacial acetic acid, HCl Beilstein Reference: 396360 Merck Reference: 14,417 MDL No.: MFCD00001213

Formel: C₁₄H₉NO₂
Molekulargewicht: 223.23 g/mol
Siedepunkt: 465
Schmelzpunkt: 252…255 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00001213
CAS-Nummer: 82-45-1
EINECS: 201-423-5
Merck Index: 13,00416

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR