4-Hydroxychinaldin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Hydroxy-2-methylquinoline

M-Clarity™ quality level = MQ200

H43806-10G H43806-50G
SIALH43806-10GEA 92.7 EUR
SIALH43806-10G SIALH43806-50G
4-Hydroxychinaldin, Sigma-Aldrich®
4-Hydroxychinaldin
Formel: C₁₀H₉NO
Siedepunkt: 313 °C (1013 hPa)
MDL Number: MFCD00006758
CAS-Nummer: 607-67-0
EINECS: 210-140-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR