3-(Hydroxymethyl)benzylalkohol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,3-Benzenedimethanol, m-Xylene-α,α'-diol, m-Phenylenedimethanol
B23417.03
B23417.06
B23417.14
B23417.03EA
32.1
EUR
B23417.03
B23417.06
B23417.14
3-(Hydroxymethyl)benzylalkohol ≥98%
3-(Hydroxymethyl)benzylalkohol
Purity: 98%
CAS No.: 626-18-6
Molecular Formula: C8H10O2
Formula Weight: 138.17
Melting Point: 56-60°
Boiling Point: 154-159°/13mm
MDL No.: MFCD00004649
Formel:
C₈H₁₀O₂ Siedepunkt: 154…159 °C (13 mmHg) Schmelzpunkt: 56…60 °C Dichte: 1.161 g/cm³ (18 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004649 CAS-Nummer: 626-18-6 EINECS: 210-934-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...