2,3-Dimethylphenylisocyanat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-Isocyanato-2,3-dimethylbenzene, 2,3-Xylyl isocyanate

L10225.06 L10225.03
L10225.06EA 99.9 EUR
L10225.06 L10225.03
2,3-Dimethylphenylisocyanat ≥99%
2,3-Dimethylphenylisocyanat

Purity: 99% CAS No.: 1591-99-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Storage Temperature: 2°C to 8°C Flash Point: 92°(197°F) Refractive Index: 1.5400 Odour: Pungent Boiling Point: 104-105°/50mm Beilstein Reference: 6313438 UN No.: 2206 MDL No.: MFCD00013851

Formel: C₉H₉NO
Siedepunkt: 104…105 °C (50 mmHg)
Dichte: 1.059
Flammpunkt: 92 °C (197 °F)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00013851
CAS-Nummer: 1591-99-7
UN-Nummer: 2206
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR