3,5-Bis(trifluormethyl)benzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11325.06
A11325.14
A11325.22
A11325.06EA
56.9
EUR
A11325.06
A11325.14
A11325.22
3,5-Bis(trifluormethyl)benzoesäure ≥98%
3,5-Bis(trifluormethyl)benzoesäure
Purity: 98%
CAS No.: 725-89-3
Molecular Formula: C9H4F6O2
Formula Weight: 258.12
Melting Point: 140-144°
Beilstein Reference: 2058600
Formel:
C₉H₄F₆O₂ Siedepunkt: 260 °C (1013 hPa) Schmelzpunkt: 140…144 °C Dichte: 1.420 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000388 CAS-Nummer: 725-89-3 EINECS: 211-970-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...