3,5-Dimethylphenylboronsäure
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethylbenzeneboronic acid, Dihydroxy(3,5-dimethylphenyl)borane, (3,5-Dimethylphenyl)metaboric acid
AC35929.1
AC35929.10
MAYBAC35929.1EA
0
EUR
MAYBAC35929.1
MAYBAC35929.10
3,5-Dimethylphenylboronsäure
3,5-Dimethylphenylboronsäure
Formel:
C₈H₁₁BO₂ |
MDL Number:
MFCD00185689 CAS-Nummer: 172975-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...