Phenolrot, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
M-Clarity™ quality level = MQ200
114529-25G
114529-5G
SIAL114529-25GEA
108
EUR
SIAL114529-25G
SIAL114529-5G
Phenolrot, Sigma-Aldrich®
Phenolrot
Formel:
C₁₉H₁₄O₅S Schmelzpunkt: > 300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003552 CAS-Nummer: 143-74-8 EINECS: 205-609-7 Merck Index: 13,07329 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...