Tetrabromphenolblau, rein
Lieferant: Thermo Fisher Scientific
Synonyme:
4,5,6,7-Tetrabromo-3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(4,5,6,7-Tetrabromo-3H-2,1-benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide
189910050
189910250
ACRO189910050EA
109
EUR
ACRO189910050
ACRO189910250
Tetrabromphenolblau, rein
Tetrabromphenolblau
Formel:
C₁₉H₆Br₈O₅S Molekulargewicht: 985.55 g/mol Schmelzpunkt: 203…205 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005876 CAS-Nummer: 4430-25-5 EINECS: 224-622-9 |
Spezifikation Testergebnisse
Appearance | Beige Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
HPLC | ≥85 % |
UV Lambda max | 608 to 618 nm |
Specific extinction E (1%/1cm) | (0.01 g/l 50% ethanol, buffer pH 5) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...