Bromphenolblau, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
M-Clarity™ quality level = MQ200
114391-25G
114391-5G
SIAL114391-25GEA
118
EUR
SIAL114391-25G
SIAL114391-5G
Bromphenolblau, Sigma-Aldrich®
Bromphenolblau
Formel:
C₁₉H₁₀Br₄O₅S Schmelzpunkt: 273 °C Dichte: 1 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005875 CAS-Nummer: 115-39-9 EINECS: 204-086-2 Merck Index: 13,01429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...