2-Methoxybenzylalkohol ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol

A14016.14 A14016.22
A14016.14EA 56.8 EUR
A14016.14 A14016.22
2-Methoxybenzylalkohol ≥99%
2-Methoxybenzylalkohol

Purity: 99% CAS No.: 612-16-8 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 110°(230°F) Refractive Index: 1.5475 Boiling Point: 248-250° Beilstein Reference: 2043673 MDL No.: MFCD00004611

Formel: C₈H₁₀O₂
Siedepunkt: 248…250 °C
Dichte: 1.12
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00004611
CAS-Nummer: 612-16-8
EINECS: 210-296-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR