2-Nitrobenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2-Nitrophenyl)methanol
M-Clarity™ quality level = MQ100
N12805-100G
N12805-25G
SIALN12805-100GEA
526
EUR
SIALN12805-100G
SIALN12805-25G
2-Nitrobenzylalkohol, Sigma-Aldrich®
2-Nitrobenzylalkohol
Formel:
C₇H₇NO₃ Siedepunkt: 270 °C (1013 hPa) Schmelzpunkt: 71…72 °C |
MDL Number:
MFCD00007186 CAS-Nummer: 612-25-9 EINECS: 210-302-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...