3-Aminobenzylalkohol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-(Hydroxymethyl)aniline, 3-(Hydroxymethyl)aniline, 3-Aminobenzyl Alcohol, (3-Aminophenyl)methanol

L07847.06 L07847.14
L07847.06EA 35.7 EUR
L07847.06 L07847.14
3-Aminobenzylalkohol ≥98%
3-Aminobenzylalkohol

Purity: 98+% CAS No.: 1877-77-6 Molecular Formula: C7H9NO Formula Weight: 123.16 Storage Temperature: 2°C to 8°C Melting Point: 91-95° Beilstein Reference: 2205844 MDL No.: MFCD00007817

Formel: C₇H₉NO
Molekulargewicht: 123.15 g/mol
Siedepunkt: 272
Schmelzpunkt: 91…95 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00007817
CAS-Nummer: 1877-77-6
EINECS: 217-514-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR