N,N-Dimethyl-p-toluidin 99%
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene
116905000 116900025
ACRO116905000EA 124 EUR
ACRO116905000 ACRO116900025
N,N-Dimethyl-p-toluidin 99%
N,N-Dimethyl-p-toluidin
Formel: C₉H₁₃N
Siedepunkt: 211 °C (760 mmHg)
Schmelzpunkt: -15 °C
Dichte: 0.93 g/cm³
Flammpunkt: 83 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008316
CAS-Nummer: 99-97-8
EINECS: 202-805-4
UN-Nummer: 2810
ADR: 6.1,III
Bestellen
Spezifikation Testergebnisse
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % Ethanol
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Free chlorine (Cl) Passes test
Lead (Pb) ≤0.05 ppm
Acid and chloride Passes test
Acetone and aldehyde Passes test