1-Butyl-4-iodbenzol
Lieferant: Thermo Scientific
Synonyme:
4-Butyliodobenzene, 1-(4-Iodophenyl)butane, 4-Butyliodbenzol, 1-n-Butyl-4-iodobenzene
TL00743.1
TL00743.10
MAYBTL00743.1EA
0
EUR
MAYBTL00743.1
MAYBTL00743.10
1-Butyl-4-iodbenzol
1-Butyl-4-iodbenzol
Formel:
C₁₀H₁₃I |
MDL Number:
MFCD00051927 CAS-Nummer: 20651-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...