3-(Trifluormethyl)thiophenol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol
BTB06567.1
BTB06567.10
BTB06567.50
MAYBBTB06567.1EA
0
EUR
MAYBBTB06567.1
MAYBBTB06567.10
MAYBBTB06567.50
3-(Trifluormethyl)thiophenol
3-(Trifluormethyl)thiophenol
Formel:
C₇H₅F₃S Siedepunkt: 161…163 °C (1013 hPa) |
MDL Number:
MFCD00041142 CAS-Nummer: 937-00-8 UN-Nummer: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...