α,α,α-Trifluor-m-toluoylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(Trifluoromethyl)benzene-1-carbonyl chloride, 3-(Trifluoromethyl)benzoyl chloride
M-Clarity™ quality level = MQ100
250279-25ML
250279-5ML
SIAL250279-25MLEA
161
EUR
SIAL250279-25ML
SIAL250279-5ML
α,α,α-Trifluor-m-toluoylchlorid, Sigma-Aldrich®
α,α,α-Trifluor-m-toluoylchlorid
Formel:
C₈H₄ClF₃O Siedepunkt: 184…186 °C (1013 hPa) Dichte: 1.4078 g/cm³ (20 °C) |
MDL Number:
MFCD00000680 CAS-Nummer: 2251-65-2 EINECS: 218-844-5 UN-Nummer: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...