α,α,α-Trifluor-4-tolualdehyd
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde
SB00720.250
SB00720.1
MAYBSB00720.250EA
0
EUR
MAYBSB00720.250
MAYBSB00720.1
α,α,α-Trifluor-4-tolualdehyd
α,α,α-Trifluor-4-tolualdehyd
Formel:
C₈H₅F₃O |
MDL Number:
MFCD00006952 CAS-Nummer: 455-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...