α,α,α-Trifluor-4-tolualdehyd ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(Trifluoromethyl)benzaldehyde, 1-Formyl-4-trifluoromethylbenzene, p-Trifluoromethylbenzaldehyde

A15276.06 A15276.14
A15276.06EA 55 EUR
A15276.06 A15276.14
α,α,α-Trifluor-4-tolualdehyd ≥97%
α,α,α-Trifluor-4-tolualdehyd

Purity: 97% CAS No.: 455-19-6 Molecular Formula: C8H5F3O Formula Weight: 174.12 Flash Point: 65°(149°F) Refractive Index: 1.4630 Melting Point: 1-2° Boiling Point: 80-81°/25mm Beilstein Reference: 1101680 MDL No.: MFCD00006952

Formel: C₈H₅F₃O
MDL Number: MFCD00006952
CAS-Nummer: 455-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR