α,α,α-Trifluor-3-tolualdehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Trifluoromethyl)benzaldehyde, Cinacalcet Impurity 23
A10463.06
A10463.14
A10463.22
A10463.06EA
62.3
EUR
A10463.06
A10463.14
A10463.22
α,α,α-Trifluor-3-tolualdehyd ≥97%
α,α,α-Trifluor-3-tolualdehyd
Purity: 97%
CAS No.: 454-89-7
Molecular Formula: C8H5F3O
Formula Weight: 174.12
Flash Point: 68°(154°F)
Refractive Index: 1.4650
Odour: Almond-like
Boiling Point: 83-86°/30mm
Beilstein Reference: 2327537
MDL No.: MFCD00003373
Formel:
C₈H₅F₃O Siedepunkt: 83…86 °C (30 mmHg) Dichte: 1.304 Flammpunkt: 68 °C (154 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003373 CAS-Nummer: 454-89-7 EINECS: 207-228-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...