4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Trifluoromethyl)benzeneboronic acid, (4-(Trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-(trifluoromethyl)phenyl)borane, (4-(Trifluoromethyl)phenyl)boronic acid

M-Clarity™ quality level = MQ100

439320-1G 439320-5G
SIAL439320-1GEA 65.7 EUR
SIAL439320-1G SIAL439320-5G
4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
4-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
MDL Number: MFCD00151855
CAS-Nummer: 128796-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR