3-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane
M-Clarity™ quality level = MQ100
432032-1G
432032-5G
SIAL432032-1GEA
53.9
EUR
SIAL432032-1G
SIAL432032-5G
3-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
3-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ |
MDL Number:
MFCD00151854 CAS-Nummer: 1423-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...