3-(Trifluormethyl)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane
B21661.03
B21661.06
B21661.14
B21661.03EA
36.8
EUR
B21661.03
B21661.06
B21661.14
3-(Trifluormethyl)phenylboronsäure ≥98%
3-(Trifluormethyl)phenylboronsäure
Purity: 98%
CAS No.: 1423-26-3
Molecular Formula: C7H6BF3O2
Formula Weight: 189.93
Melting Point: 162-166°
Beilstein Reference: 6084746
Formel:
C₇H₆BF₃O₂ Schmelzpunkt: 162…166 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151854 CAS-Nummer: 1423-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...