3,5-Dimethoxy-p-toluylsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Dimethoxy-4-methylbenzoic acid
A14801.06
A14801.14
A14801.22
A14801.06EA
42.7
EUR
A14801.06
A14801.14
A14801.22
3,5-Dimethoxy-p-toluylsäure ≥97%
3,5-Dimethoxy-p-toluylsäure
Purity: 97%
CAS No.: 61040-81-1
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 210-216°
Beilstein Reference: 2695238
Formel:
C₁₀H₁₂O₄ Schmelzpunkt: 210…216 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00017506 CAS-Nummer: 61040-81-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...