4-Methoxy-o-toluylsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H27096.06
H27096.03
H27096.06EA
139
EUR
H27096.06
H27096.03
4-Methoxy-o-toluylsäure ≥97%
4-Methoxy-o-toluylsäure
Purity: 97%
CAS No.: 6245-57-4
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 177-181°
MDL No.: MFCD00020291
Formel:
C₉H₁₀O₃ Schmelzpunkt: 177…181 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00020291 CAS-Nummer: 6245-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...