3'-Aminoacetophenon 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
m-Aminoacetophenone, 1-(3-Aminophenyl)ethanone
A12503.0B
A12503.18
A12503.30
A12503.0BEA
360
EUR
A12503.0B
A12503.18
A12503.30
3'-Aminoacetophenon 97%
3'-Aminoacetophenon
Purity: 97%
CAS No.: 99-03-6
Molecular Formula: C8H9NO
Formula Weight: 135.17
Melting Point: 94-98°
Boiling Point: 289-291°
Merck Reference: 14,413
MDL No.: MFCD00007796
Formel:
C₈H₉NO Siedepunkt: 289…291 °C Schmelzpunkt: 94…98 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007796 CAS-Nummer: 99-03-6 EINECS: 202-722-3 Merck Index: 14,413 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...