2-Acetylbenzoesäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
A12801-25G
A12801-5G
SIALA12801-25GEA
107
EUR
SIALA12801-25G
SIALA12801-5G
2-Acetylbenzoesäure, Sigma-Aldrich®
2-Acetylbenzoesäure
Formel:
C₉H₈O₃ Siedepunkt: 110…112 °C (2 mmHg) Schmelzpunkt: 112…115 °C |
MDL Number:
MFCD00002475 CAS-Nummer: 577-56-0 EINECS: 209-413-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...