ω,3-Dibromacetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Bromo-1-(3-bromophenyl)ethanone, 2-Bromo-1-(3-bromophenyl)-1-ethanone, α,3-Dibromoacetophenone, 2,3'-Dibromoacetophenone, 3-Bromophenacyl bromide

A15210.06 A15210.14
A15210.06EA 85.8 EUR
A15210.06 A15210.14
ω,3-Dibromacetophenon ≥97%
ω,3-Dibromacetophenon

Purity: 97% CAS No.: 18523-22-3 Molecular Formula: C8H6Br2O Formula Weight: 277.95 Storage Temperature: 2°C to 8°C Odour: Pungent Beilstein Reference: 775786 UN No.: 3261 MDL No.: MFCD00082698

Formel: C₈H₆Br₂O
Molekulargewicht: 277.94 g/mol
Schmelzpunkt: 49…52 °C
MDL Number: MFCD00082698
CAS-Nummer: 18523-22-3
UN-Nummer: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR