α-Brom-2-chlor-4-fluortoluol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

B23565.06 B23565.14
B23565.06EA 19.3 EUR
B23565.06 B23565.14
α-Brom-2-chlor-4-fluortoluol ≥97%
α-Brom-2-chlor-4-fluortoluol

Purity: 97% CAS No.: 45767-66-6 Molecular Formula: C7H5BrClF Formula Weight: 223.48 Melting Point: 35-39° Beilstein Reference: 3539265 UN No.: 3261 MDL No.: MFCD00236025

Formel: C₇H₅BrClF
Schmelzpunkt: 35…39 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00236025
CAS-Nummer: 45767-66-6
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR