ω-Brom-3,4-dichloracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one, 2-Bromo-1-(3,4-dichlorophenyl)ethanone, 2-Bromo-3',4'-dichloroacetophenone, 3,4-Dichlorophenacyl bromide, α-Bromo-3,4-dichloroacetophenone

663174-25G 663174-5G
SIAL663174-25GEA 198 EUR
SIAL663174-25G SIAL663174-5G
ω-Brom-3,4-dichloracetophenon, Sigma-Aldrich®
ω-Brom-3,4-dichloracetophenon
Formel: C₈H₅BrCl₂O
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00051581
CAS-Nummer: 2632-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR