1-Fluor-4-ethinylbenzol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne
M-Clarity™ quality level = MQ200
404330-1G
404330-5G
SIAL404330-1GEA
57.3
EUR
SIAL404330-1G
SIAL404330-5G
1-Fluor-4-ethinylbenzol, Sigma-Aldrich®
1-Fluor-4-ethinylbenzol
Formel:
C₈H₅F Flammpunkt: <61 °C |
MDL Number:
MFCD00168823 CAS-Nummer: 766-98-3 UN-Nummer: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...